Product Details

SNP ID
rs838102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119227157 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGCGAATCTGAGTGAGGAGACAC[A/G]GTGGCAGTAAGGCTCAGGACCTATA
Phenotype
MIM: 609671
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STEAP3 PubMed Links

Gene Details

Gene
STEAP3
Gene Name
STEAP3 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008410.1 Intron NP_001008410.1
NM_018234.2 Intron NP_060704.2
NM_138637.2 Intron NP_619543.2
NM_182915.2 Intron NP_878919.2
XM_006712614.3 Intron XP_006712677.1
XM_006712615.1 Intron XP_006712678.1
XM_011511403.1 Intron XP_011509705.1

View Full Product Details