Product Details

SNP ID
rs1446987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28990888 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCTGTCTCCTCCGCATGAGAGGC[A/G]TCTCTGTCACCTGGGTCTGTCTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM179A PubMed Links

Gene Details

Gene
FAM179A
Gene Name
family with sequence similarity 179 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321538.1 Intron NP_001308467.1
NM_001321539.1 Intron NP_001308468.1
NM_199280.2 Intron NP_954974.2
XM_011532628.2 Intron XP_011530930.1
XM_011532631.1 Intron XP_011530933.1
XM_017003507.1 Intron XP_016858996.1
XM_017003508.1 Intron XP_016858997.1
XM_017003509.1 Intron XP_016858998.1
XM_017003510.1 Intron XP_016858999.1
XM_017003511.1 Intron XP_016859000.1
XM_017003512.1 Intron XP_016859001.1
XM_017003513.1 Intron XP_016859002.1
XM_017003514.1 Intron XP_016859003.1
XM_017003515.1 Intron XP_016859004.1
XM_017003516.1 Intron XP_016859005.1

View Full Product Details