Product Details

SNP ID
rs3766183
Assay Type
Validated
NCBI dbSNP Submissions
38
Location
Chr.1:1353091 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
ATGGTGGTACAGGTGGGGGAGCTCT[T/C]GGTGGCAGGCAGCACCCCAGGAGGA
Phenotype
MIM: 601365
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DVL1 PubMed Links

Gene Details

Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
There are no transcripts associated with this gene.

Gene
MXRA8
Gene Name
matrix remodeling associated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282582.1 2132 UTR 3 NP_001269511.1
NM_001282583.1 2132 UTR 3 NP_001269512.1
NM_001282584.1 2132 UTR 3 NP_001269513.1
NM_001282585.1 2132 UTR 3 NP_001269514.1
NM_032348.3 2132 UTR 3 NP_115724.1
XM_017001516.1 2132 UTR 3 XP_016857005.1
XM_017001517.1 2132 UTR 3 XP_016857006.1

View Full Product Details