Product Details

SNP ID
rs1044258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101845957 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCTAGAATATACCAGTGCTGTT[C/T]GCTCCCGCCATCCTGAGGGCTTTAG
Phenotype
MIM: 604661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf76 PubMed Links

Gene Details

Gene
C10orf76
Gene Name
chromosome 10 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024541.2 3788 UTR 3 NP_078817.2
XM_006717969.1 3788 UTR 3 XP_006718032.1
XM_011540151.2 3788 UTR 3 XP_011538453.1
XM_011540152.2 3788 UTR 3 XP_011538454.1
XM_011540153.2 3788 UTR 3 XP_011538455.1
XM_011540154.2 3788 UTR 3 XP_011538456.1
XM_011540156.2 3788 UTR 3 XP_011538458.1
XM_011540157.2 3788 UTR 3 XP_011538459.1
XM_017016635.1 3788 Intron XP_016872124.1
XM_017016636.1 3788 Intron XP_016872125.1
Gene
KCNIP2
Gene Name
potassium voltage-gated channel interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details