Product Details

SNP ID
rs229878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74310580 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCTACTTTTGTTTGTACAGGCA[C/T]TGGTTGATGGAGATTTTACTTTTGG
Phenotype
MIM: 603432
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLIP2 PubMed Links

Gene Details

Gene
CLIP2
Gene Name
CAP-Gly domain containing linker protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003388.4 Intron NP_003379.3
NM_032421.2 Intron NP_115797.1

View Full Product Details