Product Details
- SNP ID
-
rs1046117
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:75279987 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TGGTGCAGCCCGCCCTCGTCTCCTC[C/T]GTGGCCCCATCGCAGACCAGAGCCC
- Phenotype
-
MIM: 164810
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FOS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76195634] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOS
- Gene Name
- Fos proto-oncogene, AP-1 transcription factor subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005252.3 |
457 |
Silent Mutation |
TCC,TCT |
S84S |
NP_005243.1 |
View Full Product Details