Product Details

SNP ID
rs176944
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:77709868 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTTCAAAATTTAGTCTCTGTGGATT[G/T]TTGGAAGGATTAGTGGACTTTTACT
Phenotype
MIM: 605345 MIM: 610211 MIM: 603055
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALKBH1 PubMed Links

Gene Details

Gene
ALKBH1
Gene Name
alkB homolog 1, histone H2A dioxygenase
There are no transcripts associated with this gene.

Gene
SLIRP
Gene Name
SRA stem-loop interacting RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267863.1 Intron NP_001254792.1
NM_001267864.1 Intron NP_001254793.1
NM_031210.5 Intron NP_112487.1
Gene
SNW1
Gene Name
SNW domain containing 1
There are no transcripts associated with this gene.

View Full Product Details