Product Details

SNP ID
rs1059429
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:234607782 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACAGCCACGGCTGGGCCTCTCC[A/G]TTGATCTTAGGGGGCCCGACTTCAC
Phenotype
MIM: 615332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRF2BP2 PubMed Links

Gene Details

Gene
IRF2BP2
Gene Name
interferon regulatory factor 2 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077397.1 1150 Silent Mutation AAC,AAT N357N NP_001070865.1
NM_182972.2 1150 Silent Mutation AAC,AAT N373N NP_892017.2

View Full Product Details