Product Details

SNP ID
rs438559
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151346614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAACTTCCATACAAACCCCTAA[A/G]TGACAGATTTTGGACAGCTTCCAAA
Phenotype
MIM: 607981
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NUB1 PubMed Links
Additional Information
For this assay, SNP(s) [rs115799346] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NUB1
Gene Name
negative regulator of ubiquitin like proteins 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243351.1 Intron NP_001230280.1
NM_016118.4 Intron NP_057202.3
XM_005250011.2 Intron XP_005250068.1
XM_017012304.1 Intron XP_016867793.1
XM_017012305.1 Intron XP_016867794.1
XM_017012306.1 Intron XP_016867795.1
XM_017012307.1 Intron XP_016867796.1
XM_017012308.1 Intron XP_016867797.1
XM_017012309.1 Intron XP_016867798.1
XM_017012310.1 Intron XP_016867799.1
XM_017012311.1 Intron XP_016867800.1
XM_017012312.1 Intron XP_016867801.1
XM_017012313.1 Intron XP_016867802.1
XM_017012314.1 Intron XP_016867803.1

View Full Product Details