Product Details

SNP ID
rs2070923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11275998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGGACTCTGGGTGGGGACGGGG[G/T]TTAGGGGGCTGGAGCTTTTGTCAGG
Phenotype
MIM: 182880 MIM: 182890 MIM: 190232
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRM1 PubMed Links

Gene Details

Gene
PRM1
Gene Name
protamine 1
There are no transcripts associated with this gene.

Gene
PRM2
Gene Name
protamine 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286356.1 Intron NP_001273285.1
NM_001286357.1 Intron NP_001273286.1
NM_001286358.1 Intron NP_001273287.1
NM_001286359.1 Intron NP_001273288.1
NM_002762.3 Intron NP_002753.2
Gene
PRM3
Gene Name
protamine 3
There are no transcripts associated with this gene.

Gene
TNP2
Gene Name
transition protein 2 (during histone to protamine replacement)
There are no transcripts associated with this gene.

View Full Product Details