Product Details

SNP ID
rs498079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:133241318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGCGCAAGTGCGAGGCTGCC[C/G]GCAGCGCGGCGCACGCTCCGGCCGT
Phenotype
MIM: 603550
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EYA4 PubMed Links

Gene Details

Gene
EYA4
Gene Name
EYA transcriptional coactivator and phosphatase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301012.1 Intron NP_001287941.1
NM_001301013.1 Intron NP_001287942.1
NM_004100.4 Intron NP_004091.3
NM_172103.3 Intron NP_742101.2
NM_172105.3 Intron NP_742103.1
XM_005266851.4 Intron XP_005266908.1
XM_005266853.4 Intron XP_005266910.1
XM_017010368.1 Intron XP_016865857.1
XM_017010369.1 Intron XP_016865858.1
XM_017010370.1 Intron XP_016865859.1
XM_017010371.1 Intron XP_016865860.1
XM_017010372.1 Intron XP_016865861.1
XM_017010373.1 Intron XP_016865862.1
XM_017010374.1 Intron XP_016865863.1
XM_017010375.1 Intron XP_016865864.1

View Full Product Details