Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020936.2 | 1515 | Intron | NP_065987.1 | ||
XM_011538608.2 | 1515 | Intron | XP_011536910.1 | ||
XM_017019710.1 | 1515 | Intron | XP_016875199.1 | ||
XM_017019711.1 | 1515 | Intron | XP_016875200.1 | ||
XM_017019712.1 | 1515 | Intron | XP_016875201.1 | ||
XM_017019713.1 | 1515 | Intron | XP_016875202.1 | ||
XM_017019714.1 | 1515 | Intron | XP_016875203.1 | ||
XM_017019715.1 | 1515 | Intron | XP_016875204.1 | ||
XM_017019716.1 | 1515 | Intron | XP_016875205.1 | ||
XM_017019717.1 | 1515 | Intron | XP_016875206.1 | ||
XM_017019718.1 | 1515 | Intron | XP_016875207.1 | ||
XM_017019719.1 | 1515 | Intron | XP_016875208.1 | ||
XM_017019720.1 | 1515 | Intron | XP_016875209.1 | ||
XM_017019721.1 | 1515 | Intron | XP_016875210.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001414.3 | 1515 | UTR 3 | NP_001405.1 |