Product Details

SNP ID
rs1365201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94642495 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCGCTGTGGCGTAGACGTGCAGG[C/T]GGGCCCGGCGGGTGTGCAAGAAGTA
Phenotype
MIM: 612959
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESRP1 PubMed Links

Gene Details

Gene
ESRP1
Gene Name
epithelial splicing regulatory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034915.2 Intron NP_001030087.2
NM_001122825.1 Intron NP_001116297.1
NM_001122826.1 Intron NP_001116298.1
NM_001122827.1 Intron NP_001116299.1
NM_017697.3 Intron NP_060167.2
XM_005250991.3 Intron XP_005251048.1
XM_005250992.3 Intron XP_005251049.1
Gene
LOC100288748
Gene Name
uncharacterized LOC100288748
There are no transcripts associated with this gene.

View Full Product Details