Product Details
- SNP ID
-
rs3909704
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:80488565 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAATGTTTAATGTGTAGTTATTTG[C/T]ATCAAGTAGTGGATTTGGATTGGTA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZBTB10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77448465] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZBTB10
- Gene Name
- zinc finger and BTB domain containing 10
View Full Product Details