Product Details

SNP ID
rs831989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:107823682 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATTTGTGAAAATATGTTTTTATC[A/G]TCTGCCATGTTTTGTGTTCTGTTTA
Phenotype
MIM: 612707
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GACAT1 PubMed Links

Gene Details

Gene
GACAT1
Gene Name
gastric cancer associated transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RGPD4
Gene Name
RANBP2-like and GRIP domain containing 4
There are no transcripts associated with this gene.

Gene
RGPD4-AS1
Gene Name
RGPD4 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details