Product Details

SNP ID
rs840264
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:19602110 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTGGTTTCTTTACTTGATTGTT[C/T]CTTCTCCCTGGAATGTAAACTTCAT
Phenotype
MIM: 616574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105378614 PubMed Links

Gene Details

Gene
LOC105378614
Gene Name
uncharacterized LOC105378614
There are no transcripts associated with this gene.

Gene
MINOS1
Gene Name
mitochondrial inner membrane organizing system 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032363.3 Intron NP_001027535.1
NM_001204082.1 Intron NP_001191011.1
NM_001204083.1 Intron NP_001191012.1
Gene
MINOS1-NBL1
Gene Name
MINOS1-NBL1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204088.1 Intron NP_001191017.1
NM_001204089.1 Intron NP_001191018.1
Gene
RPS14P3
Gene Name
ribosomal protein S14 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details