Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001314030.1 | 1629 | Intron | NP_001300959.1 | ||
NM_012284.2 | 1629 | Intron | NP_036416.1 | ||
XM_011538085.2 | 1629 | Intron | XP_011536387.1 | ||
XM_011538086.2 | 1629 | Intron | XP_011536388.1 | ||
XM_017019096.1 | 1629 | Intron | XP_016874585.1 | ||
XM_017019097.1 | 1629 | Intron | XP_016874586.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012300.1 | 1629 | UTR 3 | NP_001012300.1 | ||
NM_001278341.1 | 1629 | UTR 3 | NP_001265270.1 | ||
NM_006337.4 | 1629 | UTR 3 | NP_006328.2 | ||
XM_005268572.3 | 1629 | UTR 3 | XP_005268629.1 | ||
XM_011537760.2 | 1629 | UTR 3 | XP_011536062.1 | ||
XM_017018689.1 | 1629 | UTR 3 | XP_016874178.1 | ||
XM_017018690.1 | 1629 | UTR 3 | XP_016874179.1 |