Product Details

SNP ID
rs1126925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121839607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCGTGGTCAGTGTGGGCGGAGC[C/G]TTGGGGGCCGAGTCCGCTGGTGGGC
Phenotype
MIM: 609695 MIM: 611055
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HPD PubMed Links

Gene Details

Gene
HPD
Gene Name
4-hydroxyphenylpyruvate dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171993.1 1343 UTR 3 NP_001165464.1
NM_002150.2 1343 UTR 3 NP_002141.1
Gene
SETD1B
Gene Name
SET domain containing 1B
There are no transcripts associated with this gene.

View Full Product Details