Product Details

SNP ID
rs1064496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:94569292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCACTCAGCCCACGCTGAGTCTG[A/G]TGCCCACAGGTAAAGGGTAATGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR5700 PubMed Links

Gene Details

Gene
MIR5700
Gene Name
microRNA 5700
There are no transcripts associated with this gene.

Gene
MIR7844
Gene Name
microRNA 7844
There are no transcripts associated with this gene.

Gene
TMCC3
Gene Name
transmembrane and coiled-coil domain family 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301036.1 3728 UTR 3 NP_001287965.1
NM_020698.3 3728 UTR 3 NP_065749.2

View Full Product Details