Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004839.3 | 3088 | Intron | NP_004830.2 | ||
NM_199330.2 | 3088 | Intron | NP_955362.1 | ||
XM_005272448.4 | 3088 | UTR 3 | XP_005272505.1 | ||
XM_005272449.3 | 3088 | UTR 3 | XP_005272506.1 | ||
XM_006720775.3 | 3088 | Intron | XP_006720838.1 | ||
XM_006720776.3 | 3088 | UTR 3 | XP_006720839.1 | ||
XM_011522231.2 | 3088 | UTR 3 | XP_011520533.1 | ||
XM_011522232.2 | 3088 | Intron | XP_011520534.1 | ||
XM_011522233.2 | 3088 | UTR 3 | XP_011520535.1 | ||
XM_011522234.2 | 3088 | UTR 3 | XP_011520536.1 |