Product Details

SNP ID
rs1559829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49973448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGGAATGTGTTGAAAGGTGT[C/G]CATGTATGTGCGGGTGGGGTGGAGT
Phenotype
MIM: 601911
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLX4 PubMed Links

Gene Details

Gene
DLX4
Gene Name
distal-less homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001934.3 Intron NP_001925.2
NM_138281.2 Intron NP_612138.1

View Full Product Details