Product Details

SNP ID
rs942616
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:160291035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATGAGATGTGTATGTGAAGCTT[G/T]GTGGTGGTTGTGATGGGATATCTGG
Phenotype
MIM: 601924 MIM: 600279
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098398.1 Intron NP_001091868.1
NM_004371.3 Intron NP_004362.2
Gene
PEX19
Gene Name
peroxisomal biogenesis factor 19
There are no transcripts associated with this gene.

View Full Product Details