Product Details

SNP ID
rs1429967
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:40532612 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGCAGAACCCTCTTGTTGAAAGTG[C/G]AGAGCAACATTACAGTGGCTAAATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZMAT4 PubMed Links

Gene Details

Gene
ZMAT4
Gene Name
zinc finger matrin-type 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135731.1 Intron NP_001129203.1
NM_024645.2 Intron NP_078921.1
XM_017013836.1 Intron XP_016869325.1
XM_017013837.1 Intron XP_016869326.1
XM_017013838.1 Intron XP_016869327.1
XM_017013839.1 Intron XP_016869328.1
XM_017013840.1 Intron XP_016869329.1

View Full Product Details