Product Details

SNP ID
rs1362819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:24461905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAAGGGATCTATAGAAGATTAG[A/G]TTAGTCTCTTCCAGTAAGTTTTTGA
Phenotype
MIM: 606792 MIM: 615319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRH4 PubMed Links

Gene Details

Gene
HRH4
Gene Name
histamine receptor H4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143828.1 Intron NP_001137300.1
NM_001160166.1 Intron NP_001153638.1
NM_021624.3 Intron NP_067637.2
XM_011526133.1 Intron XP_011524435.1
Gene
IMPACT
Gene Name
impact RWD domain protein
There are no transcripts associated with this gene.

View Full Product Details