Product Details
- SNP ID
-
rs8810
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:16327504 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTGCTGGGTTGGTTTTTTTTTT[A/T]ATTAAAAAGTTTTGCATCTTTTAAA
- Phenotype
-
MIM: 602016
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLF2
PubMed Links
Gene Details
- Gene
- KLF2
- Gene Name
- Kruppel like factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016270.3 |
1639 |
UTR 3 |
|
|
NP_057354.1 |
View Full Product Details