Product Details

SNP ID
rs1793167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118231343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGTTTTCTTCCTATCTCACTAGC[T/C]GCTCTTTCTTAGCCTCTTTACTAAT
Phenotype
MIM: 609770 MIM: 611707
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
JAML PubMed Links

Gene Details

Gene
JAML
Gene Name
junction adhesion molecule like
There are no transcripts associated with this gene.

Gene
MPZL3
Gene Name
myelin protein zero like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286152.1 Intron NP_001273081.1
NM_198275.2 Intron NP_938016.1
XM_006718775.2 Intron XP_006718838.1
XM_017017304.1 Intron XP_016872793.1

View Full Product Details