Product Details

SNP ID
rs941726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102783339 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGGGTTCTTTGATGGAAAGTTT[C/T]ATTTGACAATGTAATTTTCTTGGAT
Phenotype
MIM: 601896
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAF3 PubMed Links

Gene Details

Gene
TRAF3
Gene Name
TNF receptor associated factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199427.1 Intron NP_001186356.1
NM_003300.3 Intron NP_003291.2
NM_145725.2 Intron NP_663777.1
NM_145726.2 Intron NP_663778.1
XM_011537116.2 Intron XP_011535418.1
XM_011537117.2 Intron XP_011535419.1
XM_011537118.2 Intron XP_011535420.1
XM_017021617.1 Intron XP_016877106.1
XM_017021618.1 Intron XP_016877107.1
XM_017021619.1 Intron XP_016877108.1
XM_017021620.1 Intron XP_016877109.1

View Full Product Details