Product Details

SNP ID
rs652303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60202574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAACTGAGAGCTATTACCCAAAC[C/T]GAAAACAGTCCTAGTTTCCAAATAT
Phenotype
MIM: 608401
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6503 PubMed Links

Gene Details

Gene
MIR6503
Gene Name
microRNA 6503
There are no transcripts associated with this gene.

Gene
MS4A4E
Gene Name
membrane spanning 4-domains A4E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011545416.2 Intron XP_011543718.1
XM_011545417.2 Intron XP_011543719.1
XM_011545418.2 Intron XP_011543720.1
XM_011545419.2 Intron XP_011543721.1
XM_017018641.1 Intron XP_016874130.1

View Full Product Details