Product Details

SNP ID
rs1618462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:107594996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGAAGACCCAGAAGCTACTTTC[A/G]TTATTGAATAACGTGGTAAATTTCC
Phenotype
MIM: 615456
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELMOD1 PubMed Links

Gene Details

Gene
ELMOD1
Gene Name
ELMO domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130037.1 Intron NP_001123509.1
NM_001308018.1 Intron NP_001294947.1
NM_018712.3 Intron NP_061182.3
XM_017017994.1 Intron XP_016873483.1
XM_017017995.1 Intron XP_016873484.1
Gene
LOC643923
Gene Name
uncharacterized LOC643923
There are no transcripts associated with this gene.

View Full Product Details