Product Details

SNP ID
rs860693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:44933830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGCAGCCCGGGCTGCACGTGT[C/G]GGGGGCAGCTCACCGGGAAGCAGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TP53I11 PubMed Links

Gene Details

Gene
TP53I11
Gene Name
tumor protein p53 inducible protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258320.1 2085 UTR 3 NP_001245249.1
NM_001258321.1 2085 UTR 3 NP_001245250.1
NM_001258322.1 2085 UTR 3 NP_001245251.1
NM_001258323.2 2085 UTR 3 NP_001245252.1
NM_001258324.1 2085 UTR 3 NP_001245253.1
NM_001318384.1 2085 UTR 3 NP_001305313.1
NM_001318385.1 2085 UTR 3 NP_001305314.1
NM_001318386.1 2085 UTR 3 NP_001305315.1
NM_001318387.1 2085 UTR 3 NP_001305316.1
NM_001318388.1 2085 UTR 3 NP_001305317.1
NM_001318389.1 2085 UTR 3 NP_001305318.1
NM_001318390.1 2085 UTR 3 NP_001305319.1
NM_006034.4 2085 UTR 3 NP_006025.2
XM_005253227.4 2085 UTR 3 XP_005253284.1
XM_005253229.1 2085 Intron XP_005253286.1
XM_011520475.2 2085 UTR 3 XP_011518777.1
XM_011520476.2 2085 UTR 3 XP_011518778.1
XM_011520477.2 2085 UTR 3 XP_011518779.1
XM_011520478.2 2085 UTR 3 XP_011518780.1
XM_017018580.1 2085 UTR 3 XP_016874069.1
XM_017018581.1 2085 UTR 3 XP_016874070.1
Gene
TSPAN18
Gene Name
tetraspanin 18
There are no transcripts associated with this gene.

View Full Product Details