Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001119.4 | 1558 | Intron | NP_001110.2 | ||
NM_001286645.1 | 1558 | Intron | NP_001273574.1 | ||
NM_014189.3 | 1558 | Intron | NP_054908.2 | ||
NM_014190.3 | 1558 | Intron | NP_054909.2 | ||
NM_176801.2 | 1558 | Intron | NP_789771.1 | ||
XM_005247933.2 | 1558 | Intron | XP_005247990.1 | ||
XM_005247934.2 | 1558 | Intron | XP_005247991.1 | ||
XM_005247935.2 | 1558 | Intron | XP_005247992.1 | ||
XM_005247936.2 | 1558 | Intron | XP_005247993.1 | ||
XM_005247937.2 | 1558 | Intron | XP_005247994.1 | ||
XM_017007703.1 | 1558 | Intron | XP_016863192.1 | ||
XM_017007704.1 | 1558 | Intron | XP_016863193.1 | ||
XM_017007705.1 | 1558 | Intron | XP_016863194.1 | ||
XM_017007706.1 | 1558 | Intron | XP_016863195.1 | ||
XM_017007707.1 | 1558 | Intron | XP_016863196.1 | ||
XM_017007708.1 | 1558 | Intron | XP_016863197.1 | ||
XM_017007709.1 | 1558 | Intron | XP_016863198.1 | ||
XM_017007710.1 | 1558 | Intron | XP_016863199.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001120.4 | 1558 | Silent Mutation | NP_001111.3 | ||
NM_001146069.1 | 1558 | Silent Mutation | NP_001139541.1 | ||
XM_005247927.3 | 1558 | UTR 3 | XP_005247984.1 |