Product Details

SNP ID
rs1801800
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:46675250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGCTCAAATCAAGTCATGGAA[A/G]CCCTGAATTCTCCCTAAACAGAACC
Phenotype
MIM: 608917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
There are no transcripts associated with this gene.

Gene
EFCAB14
Gene Name
EF-hand calcium binding domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014774.2 5676 UTR 3 NP_055589.1
Gene
EFCAB14-AS1
Gene Name
EFCAB14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TEX38
Gene Name
testis expressed 38
There are no transcripts associated with this gene.

View Full Product Details