Product Details

SNP ID
rs1054130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37523204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCCTGAATCTTGGTGCAGCACC[A/C]ACCGCGTGCCTGTGTGGCGGGACTG
Phenotype
MIM: 613110 MIM: 603106
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BLCAP PubMed Links

Gene Details

Gene
BLCAP
Gene Name
bladder cancer associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167820.1 736 Intron NP_001161292.1
NM_001167821.1 736 Intron NP_001161293.1
NM_001167822.2 736 Intron NP_001161294.1
NM_001167823.1 736 Intron NP_001161295.1
NM_001317074.1 736 Intron NP_001304003.1
NM_001317075.1 736 Intron NP_001304004.1
NM_006698.3 736 Intron NP_006689.1
XM_017027599.1 736 Intron XP_016883088.1
Gene
NNAT
Gene Name
neuronatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322802.1 736 UTR 3 NP_001309731.1
NM_005386.3 736 UTR 3 NP_005377.1
NM_181689.2 736 UTR 3 NP_859017.1

View Full Product Details