Product Details

SNP ID
rs911933
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:98944892 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGCGGTTGGCTGAATAGTAAGATA[C/T]AAGTTAAAGTAGACCTAAGTTAAAA
Phenotype
MIM: 120325
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL15A1 PubMed Links

Gene Details

Gene
COL15A1
Gene Name
collagen type XV alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001855.4 Intron NP_001846.3
XM_011518214.2 Intron XP_011516516.1

View Full Product Details