Product Details

SNP ID
rs943591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55271875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCTTGCACCGGACCGGCGCTCC[A/C]ACTTACGGGACCCAGGGCGGTGAGG
Phenotype
MIM: 609104
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FBXO34 PubMed Links

Gene Details

Gene
FBXO34
Gene Name
F-box protein 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017943.3 Intron NP_060413.2
NM_152231.1 Intron NP_689417.1
XM_005267791.4 Intron XP_005267848.1
XM_006720185.3 Intron XP_006720248.1
XM_017021391.1 Intron XP_016876880.1
XM_017021392.1 Intron XP_016876881.1
XM_017021393.1 Intron XP_016876882.1

View Full Product Details