Product Details

SNP ID
rs913485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60974470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCAGATGATATGTAAACAAA[C/T]GAGCATGGTATGTTCCAATCAAACT
Phenotype
MIM: 602659 MIM: 616518 MIM: 611023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MNAT1 PubMed Links

Gene Details

Gene
MNAT1
Gene Name
MNAT1, CDK activating kinase assembly factor
There are no transcripts associated with this gene.

Gene
SLC38A6
Gene Name
solute carrier family 38 member 6
There are no transcripts associated with this gene.

Gene
TRMT5
Gene Name
tRNA methyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020810.3 2275 UTR 3 NP_065861.3
XM_005267916.3 2275 UTR 3 XP_005267973.2
XM_011537017.2 2275 Intron XP_011535319.1

View Full Product Details