Product Details
- SNP ID
-
rs963393
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:144681744 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAGGTAGTCATGTAAAAATTAAA[A/G]AGAAATAATTTATAAAGTCTTTGTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TEX41
PubMed Links
Gene Details
- Gene
- TEX41
- Gene Name
- testis expressed 41 (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details