Product Details

SNP ID
rs809418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35775920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGCTGGGAGGAGGCGGGGCT[A/G]GTCCAGGAAGGGACCCGCGCCACCC
Phenotype
MIM: 614902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP33 PubMed Links

Gene Details

Gene
ARHGAP33
Gene Name
Rho GTPase activating protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172630.1 Intron NP_001166101.1
NM_052948.3 Intron NP_443180.2
XM_005258488.1 Intron XP_005258545.1
XM_006722999.1 Intron XP_006723062.1
XM_006723003.2 Intron XP_006723066.1
XM_011526416.1 Intron XP_011524718.1
XM_011526417.1 Intron XP_011524719.1
XM_011526418.1 Intron XP_011524720.1
XM_011526419.1 Intron XP_011524721.1
XM_011526420.1 Intron XP_011524722.1
XM_011526421.2 Intron XP_011524723.1
XM_017026238.1 Intron XP_016881727.1
Gene
PROSER3
Gene Name
proline and serine rich 3
There are no transcripts associated with this gene.

View Full Product Details