Product Details
- SNP ID
-
rs1801695
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:21001981 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAATTATTTTCTTCGTCGCAATGG[C/T]CTGGCTTTTAATTATTTCCTGAGCA
- Phenotype
-
MIM: 107730
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOB
PubMed Links
Gene Details
- Gene
- APOB
- Gene Name
- apolipoprotein B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000384.2 |
13569 |
Missense Mutation |
|
|
NP_000375.2 |
View Full Product Details