Product Details

SNP ID
rs1466864
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:85100004 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CGGGCAGCCCTCCATGCTCCCACCA[A/C]CCCTGCCTCCTCAGCCTCCCATTTC
Phenotype
MIM: 611675
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM92B PubMed Links

Gene Details

Gene
FAM92B
Gene Name
family with sequence similarity 92 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198491.2 Intron NP_940893.1
XM_005255931.2 Intron XP_005255988.1
XM_011523063.1 Intron XP_011521365.1
XM_017023198.1 Intron XP_016878687.1
Gene
KIAA0513
Gene Name
KIAA0513
There are no transcripts associated with this gene.

View Full Product Details