Product Details

SNP ID
rs1802381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:83809394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTGGAAGAACTGGAAAGTGAA[A/C]ACAAGATACCTGCCACGCAAAAGAG
Phenotype
MIM: 604553
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSBP1 PubMed Links

Gene Details

Gene
HSBP1
Gene Name
heat shock factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001537.3 376 Missense Mutation AAC,CAC N68H NP_001528.1
Gene
LOC102724163
Gene Name
uncharacterized LOC102724163
There are no transcripts associated with this gene.

View Full Product Details