Product Details
- SNP ID
-
rs11721955
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:159125413 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATTCCGTTTTCTTTGTAATGTGCT[A/G]TGGAGTTTTTTTGAGTAGCTGCTAC
- Phenotype
-
MIM: 609530
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3688-1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116407841] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR3688-1
- Gene Name
- microRNA 3688-1
There are no transcripts associated with this gene.
- Gene
- MIR3688-2
- Gene Name
- microRNA 3688-2
There are no transcripts associated with this gene.
- Gene
- RAPGEF2
- Gene Name
- Rap guanine nucleotide exchange factor 2
View Full Product Details