Product Details

SNP ID
rs11820526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9677715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGAAATGAACTCATTGTCTGTG[C/T]ATTTTTATCCTGTTGTTTTTAGAAA
Phenotype
MIM: 604762
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SWAP70 PubMed Links

Gene Details

Gene
SWAP70
Gene Name
SWAP switching B-cell complex 70kDa subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297714.1 Intron NP_001284643.1
NM_015055.3 Intron NP_055870.2
XM_005252830.4 Intron XP_005252887.1

View Full Product Details