Product Details
- SNP ID
-
rs6552879
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:185401483 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GATCCAGATTTTTTAAAGGAGAAAT[G/T]GGTAGAGAGGGTGTCAGAATGGGAT
- Phenotype
-
MIM: 611482
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD37
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78882169] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD37
- Gene Name
- ankyrin repeat domain 37
There are no transcripts associated with this gene.
- Gene
- C4orf47
- Gene Name
- chromosome 4 open reading frame 47
There are no transcripts associated with this gene.
- Gene
- LRP2BP
- Gene Name
- LRP2 binding protein
There are no transcripts associated with this gene.
- Gene
- UFSP2
- Gene Name
- UFM1 specific peptidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018359.3 |
|
Intron |
|
|
NP_060829.2 |
View Full Product Details