Product Details
- SNP ID
-
rs12501580
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:185020047 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAACTGGTTCCCAGGGAGGCTGG[G/T]TCCACCTCCTTCTCCCGGGTGGGGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HELT
PubMed Links
Gene Details
- Gene
- HELT
- Gene Name
- helt bHLH transcription factor
View Full Product Details