Product Details
- SNP ID
-
rs11665792
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53608356 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGTGCAGATCTCCCTTGATGAAGCC[A/G]GAGAAGATGACGCAGCTCAGAACCA
- Phenotype
-
MIM: 611165
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DPRX
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12608484] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DPRX
- Gene Name
- divergent-paired related homeobox
- Gene
- LOC284379
- Gene Name
- solute carrier family 7 member 3 pseudogene
There are no transcripts associated with this gene.
View Full Product Details