Product Details

SNP ID
rs35710536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38303728 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCACAGAGGTAAAACAACTTTGC[C/T]GAGGTCACCTTGCTGCTAAGTGACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf33 PubMed Links

Gene Details

Gene
C19orf33
Gene Name
chromosome 19 open reading frame 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317801.1 2600 Intron NP_001304730.1
NM_033520.2 2600 Intron NP_277055.1
Gene
YIF1B
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039671.2 2600 UTR 3 NP_001034760.1
NM_001039672.2 2600 UTR 3 NP_001034761.1
NM_001039673.2 2600 UTR 3 NP_001034762.1
NM_001145461.1 2600 UTR 3 NP_001138933.1
NM_001145462.1 2600 UTR 3 NP_001138934.1
NM_001145463.1 2600 UTR 3 NP_001138935.1
XM_005259385.3 2600 Intron XP_005259442.1
XM_017027449.1 2600 Intron XP_016882938.1
XM_017027450.1 2600 Intron XP_016882939.1
XM_017027451.1 2600 Intron XP_016882940.1

View Full Product Details