Product Details

SNP ID
rs13196394
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:70673318 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCTATGGAGCTGCAACCACTTAGT[A/G]TCTGTAGACGTTTTTTGTTAGTTGC
Phenotype
MIM: 611372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMAP1 PubMed Links

Gene Details

Gene
SMAP1
Gene Name
small ArfGAP 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044305.2 Intron NP_001037770.1
NM_001281439.1 Intron NP_001268368.1
NM_001281440.1 Intron NP_001268369.1
NM_021940.4 Intron NP_068759.2
XM_005248760.4 Intron XP_005248817.1
XM_011536045.2 Intron XP_011534347.1
XM_011536046.2 Intron XP_011534348.1
XM_011536047.2 Intron XP_011534349.1
XM_011536048.2 Intron XP_011534350.1
XM_017011170.1 Intron XP_016866659.1

View Full Product Details