Product Details

SNP ID
rs1340381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80340998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACTTTTTGATTTAATTCACCAT[C/T]CACACTTTTTAAATATTGCTTTAAT
Phenotype
MIM: 615154
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYDC1 PubMed Links

Gene Details

Gene
DYDC1
Gene Name
DPY30 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001269053.1 Intron NP_001255982.1
NM_138812.3 Intron NP_620167.1
XM_005269549.4 Intron XP_005269606.1
XM_005269550.4 Intron XP_005269607.1
XM_011539334.2 Intron XP_011537636.1
XM_011539335.1 Intron XP_011537637.1
XM_017015749.1 Intron XP_016871238.1
Gene
DYDC2
Gene Name
DPY30 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details