Product Details

SNP ID
rs12631989
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:49236563 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TAAATTTCAGGAACAAGAAGTCATC[C/T]AACTGGAATAATAATCAAAATGATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC36 PubMed Links

Gene Details

Gene
CCDC36
Gene Name
coiled-coil domain containing 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135197.1 170 Silent Mutation TCC,TCT S24S NP_001128669.1
NM_178173.3 170 Silent Mutation TCC,TCT S24S NP_835467.2
XM_006713124.3 170 Silent Mutation TCC,TCT S24S XP_006713187.1
XM_011533670.2 170 Silent Mutation TCC,TCT S31S XP_011531972.1
XM_011533671.2 170 Silent Mutation TCC,TCT S31S XP_011531973.1
XM_011533672.2 170 Silent Mutation TCC,TCT S24S XP_011531974.1
XM_011533673.2 170 Silent Mutation TCC,TCT S24S XP_011531975.1

View Full Product Details